missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LRH-1/NR5A2 Antibody (PCRP-NR5A2-1B8) [Alexa Fluor™ 405], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-26934AF405

Product Code. 30065878

  • £514.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LRH-1/NR5A2 Monoclonal antibody specifically detects LRH-1/NR5A2 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRH-1/NR5A2
Monoclonal
Alexa Fluor 405
Alpha-1-fetoprotein transcription factor, B1-binding factor, b1-binding factor, hepatocyte transcription factor which activates enhancer IIof hepatitis B virus, B1F, B1F2, fetoprotein-alpha 1 (AFP) transcription factor, FTFLRH1, FTZ-F1, FTZ-F1beta, HB1F, hB1F-2, hB1FCYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, liver receptor homolog-1, LRH-1CPF, nuclear receptor NR5A2, nuclear receptor subfamily 5 group A member 2, nuclear receptor subfamily 5, group A, member 2
Recombinant human LRH-1/NR5A2 protein
0.1 mL
Cancer, Chromatin Research, Neuroscience, Stem Cell Markers
2494
Store at 4°C in the dark.
IgG2b
Flow Cytometry, Immunohistochemistry (Paraffin)
PCRP-NR5A2-1B8
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.