missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRH-1/NR5A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£177.00 - £332.00
Specifications
| Antigen | LRH-1/NR5A2 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1-5ug/ml∼, Immunohistochemistry, Immunohistochemistry-Paraffin 5ug/ml, KnockDown Validated reported in scientific literature (PMID 31978970) |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18190864
|
Novus Biologicals
NBP2-27196SS |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18791054
|
Novus Biologicals
NBP2-27196 |
0.1 mg |
£332.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRH-1/NR5A2 Polyclonal specifically detects LRH-1/NR5A2 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Chinese Hamster, Equine, Guinea Pig, Opossum, Primate, Rabbit, Rhesus Macaque, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| LRH-1/NR5A2 | |
| Western Blot 1-5ug/ml∼, Immunohistochemistry, Immunohistochemistry-Paraffin 5ug/ml, KnockDown Validated reported in scientific literature (PMID 31978970) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Chinese Hamster, Equine, Guinea Pig, Opossum, Primate, Rabbit | |
| Alpha-1-fetoprotein transcription factor, B1-binding factor, b1-binding factor, hepatocyte transcription factor which activates enhancer IIof hepatitis B virus, B1F, B1F2, fetoprotein-alpha 1 (AFP) transcription factor, FTFLRH1, FTZ-F1, FTZ-F1beta, HB1F, hB1F-2, hB1FCYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, liver receptor homolog-1, LRH-1CPF, nuclear receptor NR5A2, nuclear receptor subfamily 5 group A member 2, nuclear receptor subfamily 5, group A, member 2 | |
| NR5A2 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for this NR5A2 antibody is 100% homologous in human, mouse, rat, rhesus monkey, orangutan, chimpanzee, new world monkey, pig, panda, dog, Chinese hamster, horse, cow, african bush elephant, sheep, guinea pig, Carolina anole, zebrafish, chicken, great ape, mole rat, opossum, white-cheeked gibbon, platypus, rabbit, zebra finch, frog, and catfish. |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, Neuroscience | |
| PBS with 0.05% Sodium Azide | |
| 2494 | |
| A portion of amino acids 140-190 from human NR5A2 was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title