missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRG1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13224-1-AP-150UL
This item is not returnable.
View return policy
Description
The leucine-rich repeat (LRR) family of proteins, including LRG1, have been shown to be involved in protein-protein interaction, signal transduction, and cell adhesion and development. LRG1 is expressed during granulocyte differentiation.Specifications
| LRG1 | |
| Polyclonal | |
| Unconjugated | |
| LRG1 | |
| HMFT1766, LRG, LRG1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116844, 76905 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02750 | |
| Lrg1 | |
| LRG1 Fusion Protein Ag3964 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction