missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRG1 Monoclonal Antibody (D2)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA541628

 View more versions of this product

Product Code. 17816832

  • £430.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LRG1 Monoclonal Antibody for Western Blot, ICC/IF, IHC

The leucine-rich repeat (LRR) family of proteins, including LRG1, have been shown to be involved in protein-protein interaction, signal transduction, and cell adhesion and development. LRG1 is expressed during granulocyte differentiation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRG1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P02750
LRG1
Recombinant protein Leucine Rich Alpha-2-Glycoprotein 1. The antigen corresponds to amino acid range 37-340 of the target protein.
200 μL
Primary
Human
Antibody
IgG2a κ
Immunohistochemistry, Western Blot, Immunocytochemistry
D2
Unconjugated
LRG1
1300008B03Rik; 2310031E04Rik; A2GL; HMFT1766; leucine rich alpha 2 glycoprotein; leucine rich alpha-2-glycoprotein 1; leucine-rich alpha-2-glycoprotein; leucine-rich alpha-2-glycoprotein 1; Leucine-rich alpha-2-glycoprotein precursor; LRG; LRG 1; LRG1; LRG-1; Lrhg
Mouse
Protein A/G
RUO
116844
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.