missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRG1 Antibody (2F5.A2), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-51830
This item is not returnable.
View return policy
Description
LRG1 Monoclonal specifically detects LRG1 in Human samples. It is validated for Immunoassay.
Specifications
| LRG1 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Immunoassay 1 ug/ml | |
| 1300008B03Rik, 2310031E04Rik, HMFT1766, leucine-rich alpha-2-glycoprotein, leucine-rich alpha-2-glycoprotein 1, LRGleucine rich alpha 2 glycoprotein | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoassay | |
| 2F5.A2 | |
| Unconjugated | |
| P02750 | |
| LRG1 | |
| Human LRG, covalently coupled to hemocyanin using glutaraldehyde | |
| 0.05 mg | |
| Cancer | |
| 116844 | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction