missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRG1 Antibody (001), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90217H
This item is not returnable.
View return policy
Description
LRG1 Monoclonal specifically detects LRG1 in Human samples. It is validated for ELISA.
Specifications
| LRG1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1 | |
| ELISA | |
| 1300008B03Rik, 2310031E04Rik, HMFT1766, leucine-rich alpha-2-glycoprotein, leucine-rich alpha-2-glycoprotein 1, LRGleucine rich alpha 2 glycoprotein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human LRG1 (AAH34389.1; Met1-Gln347). | |
| 0.1 mL | |
| Cancer, Phospho Specific | |
| 116844 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction