missing translation for 'onlineSavingsMsg'
Learn More

LRG1 Antibody (001), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18335541 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335541 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335541 Supplier Novus Biologicals Supplier No. NBP290217AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

LRG1 Monoclonal specifically detects LRG1 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRG1
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 594
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias 1300008B03Rik, 2310031E04Rik, HMFT1766, leucine-rich alpha-2-glycoprotein, leucine-rich alpha-2-glycoprotein 1, LRGleucine rich alpha 2 glycoprotein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human LRG1 (AAH34389.1; Met1-Gln347).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 116844
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.