missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRFN3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12818852
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12818852 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12818852 Supplier Invitrogen™ Supplier No. PA520706

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is K562 cell lysate. PA5-20706 can be used with blocking peptide PEP-0821.

LRFN3 is one of a family of five transmembrane glycoproteins that are highly expressed in neuronal tissues. LRFN proteins share leucine-rich repeat (LRR)-immunoglobulin-like (Ig)-fibronectin type III (Fn)-transmembrane domain structure with other members of the LRR-Ig-Fn protein superfamily such as the Slitrk family of proteins. Expression of LRFN1, -3, and -4 mRNA was detected in embryonic neuronal cells, while Lrfn2 and Lrfn5 expression was primarily restricted to more mature cells. LRFN1, -2, and -4 bound to PDZ domains of postsynaptic PSD95, re-distributing PSD95 to the cell periphery. It has been suggested that the Lrfn proteins play a role in the developing and/or mature vertebrate nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRFN3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene LRFN3
Gene Accession No. B0BNK7, Q8BLY3, Q9BTN0
Gene Alias A530045B06Rik; EMBL:AAI58863.1, ECO:0000312; fibronectin type III, immunoglobulin and leucine rich repeat domains 1; FIGLER1; leucine rich repeat and fibronectin type III domain containing 3; leucine-rich repeat and fibronectin type-III domain-containing protein 3; LRFN3; lrfn3 {ECO:0000312; PubMed:18227064}; RGD:1305567}; Salm4; Synaptic adhesion-like molecule 4; synaptic adhesion-like molecule 4 {ECO:0000303; UNQ5865/PRO34192
Gene Symbols LRFN3
Host Species Rabbit
Immunogen A 16 amino acid peptide near the center of human LRFN3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233067, 308495, 79414
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.