missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRFN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520705
This item is not returnable.
View return policy
Description
A suggested positive control is rat brain tissue lysate. PA5-20705 can be used with blocking peptide PEP-0820.
LRFN2 is one of a family of five transmembrane glycoproteins that are highly expressed in neuronal tissues. LRFN proteins share leucine-rich repeat (LRR)-immunoglobulin-like (Ig)-fibronectin type III (Fn)-transmembrane domain structure with other members of the LRR-Ig-Fn protein superfamily such as the Slitrk family of proteins. Expression of LRFN1, -3, and -4 mRNA was detected in embryonic neuronal cells, while Lrfn2 and Lrfn5 expression was primarily restricted to more mature cells. LRFN1, -2, and -4 bound to PDZ domains of postsynaptic PSD95, re-distributing PSD95 to the cell periphery. It has been suggested that the Lrfn proteins play a role in the developing and/or mature vertebrate nervous system.
Specifications
| LRFN2 | |
| Polyclonal | |
| Unconjugated | |
| LRFN2 | |
| 5730420O05Rik; fibronectin type III, immunoglobulin and leucine rich repeat domains 2; FIGLER2; KIAA1246; leucine rich repeat and fibronectin type III domain containing 2; leucine-rich repeat and fibronectin type-III domain-containing protein 2; Lrfn2; mKIAA1246; RP11-535K1.2; SALM1; synaptic adhesion-like molecule 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 316205, 57497, 70530 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q460M5, Q80TG9, Q9ULH4 | |
| LRFN2 | |
| A 14 amino acid peptide near the carboxy terminus of human LRFN2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction