missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRFN1 Monoclonal Antibody (610712)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561578

Product Code. 30534565

  • £315.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

LRFN1 is one of a family of five transmembrane glycoproteins that are highly expressed in neuronal tissues. LRFN proteins share leucine-rich repeat (LRR)-immunoglobulin-like (Ig)-fibronectin type III (Fn)-transmembrane domain structure with other members of the LRR-Ig-Fn protein superfamily such as the Slitrk family of proteins. Expression of LRFN1, -3, and -4 mRNA was detected in embryonic neuronal cells, while Lrfn2 and Lrfn5 expression was primarily restricted to more mature cells. LRFN1, -2, and -4 bound to PDZ domains of postsynaptic PSD95, re-distributing PSD95 to the cell periphery. It has been suggested that the Lrfn proteins play a role in the developing and/or mature vertebrate nervous system. At least two isoforms of LRFN1 are known to exist.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRFN1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q9P244
LRFN1
Chinese hamster ovary cell line CHO-derived recombinant human SALM2, Gln32-Gly534 (predicted).
100 μg
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin)
610712
Unconjugated
LRFN1
BC004018; CTC-246B18.8; KIAA1484; leucine rich repeat and fibronectin type III domain containing 1; leucine-rich repeat and fibronectin type III domain-containing protein 1; Lrfn1; Salm2; Semo1; Synaptic adhesion-like molecule 2; synaptic differentiation enhancing molecule 1; synaptic differentiation-enhancing molecule 1
Mouse
Protein A/G
RUO
57622
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.