missing translation for 'onlineSavingsMsg'
Learn More

LRE1, MedChemExpress

Product Code. 16475797
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475797 5 mg 5mg
16423068 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16475797 Supplier Medchem Express Supplier No. HY100524

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LRE1 is a specific and allosteric inhibitor of soluble adenylyl cyclase.

LRE1 is a specific and allosteric inhibitor of soluble adenylyl cyclase.

Specifications

Chemical Name or Material LRE1
Physical Form Powder
CAS 1252362-53-0
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C12H13ClN4S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 125 mg/mL (445.19 mM; Need ultrasonic)
SMILES NC1=NC(N(CC2=CC=CS2)C3CC3)=CC(Cl)=N1
Molecular Weight (g/mol) 280.78
Formula Weight 280.78
Percent Purity 99.59%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.