missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRCH1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44281AF532
This item is not returnable.
View return policy
Description
LRCH1 Polyclonal antibody specifically detects LRCH1 in Human samples. It is validated for Western Blot
Specifications
| LRCH1 | |
| Alexa Fluor 532 | |
| CHDC1, FLJ41660, KIAA1016, leucine-rich repeat and calponin homology domain-containing protein 1, leucine-rich repeats and calponin homology (CH) domain containing 1, neuronal protein 81, NP81 | |
| Immunogen affinity purified | |
| RUO | |
| Human |
| Western Blot | |
| Western Blot | |
| Rabbit | |
| 0.1 mL | |
| 23143 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction