missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£166.00 - £376.00
Specifications
| Antigen | LRAT |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18321016
|
Novus Biologicals
NBP3-04496-20UL |
20 μL |
£166.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18017094
|
Novus Biologicals
NBP3-04496-100UL |
100 μL |
£376.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRAT Polyclonal specifically detects LRAT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| LRAT | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 9227 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| EC 2.3.1.135, LCA14, lecithin retinol acyltransferase, lecithin retinol acyltransferase (phosphatidylcholine--retinolO-acyltransferase), MGC33103, Phosphatidylcholine--retinol O-acyltransferase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-194 of human LRAT (NP_004735.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title