missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRAT Antibody (M34-P1F10), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50444AF488
This item is not returnable.
View return policy
Description
LRAT Monoclonal specifically detects LRAT in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| LRAT | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LRAT | |
| Peptide Sequence - RDQRSVLASA (amino acids 190 -199) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| M34-P1F10 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| EC 2.3.1.135, LCA14, lecithin retinol acyltransferase, lecithin retinol acyltransferase (phosphatidylcholine--retinolO-acyltransferase), MGC33103, Phosphatidylcholine--retinol O-acyltransferase | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 9227 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction