missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LR8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£390.00
Specifications
| Antigen | LR8 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
LR8 Polyclonal specifically detects LR8 in Human samples. It is validated for Western Blot.Specifications
| LR8 | |
| Polyclonal | |
| Rabbit | |
| NP_054739 | |
| 28959 | |
| Synthetic peptide directed towards the C terminal of human TMEM176B. Peptide sequence SLVSLGVGLRNLCGQSSQPLNEEGSEKRLLGENSVPPSPSREQTSTAIVL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| LR8FLJ26014, LR8-like protein, MGC110857, Protein LR8, transmembrane protein 176B | |
| TMEM176B | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title