missing translation for 'onlineSavingsMsg'
Learn More

LR-90, MedChemExpress

Product Code. 16487946
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487946 100 mg 100mg
16454107 50 mg 50mg
16448208 10 mg 10mg
16430648 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16487946 Supplier Medchem Express Supplier No. HY76383
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress LR-90 is an advanced glycation end product (AGE) inhibitor, inhibits inflammatory responses in human monocytes. LR-90 is also used in the research of diabetic animal model.

LR-90 is an advanced glycation end product (AGE) inhibitor, inhibits inflammatory responses in human monocytes. LR-90 is also used in the research of diabetic animal model.

Specifications

Chemical Name or Material LR-90
CAS 245075-84-7
Color White
Purity Grade Notes Research
Molecular Formula C35H34Cl2N4O8
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (140.93 mM)
SMILES O=C(NC1=CC=C(C=C1)OC(C)(C)C(O)=O)NC(C(Cl)=C2)=CC=C2CC3=CC=C(C(Cl)=C3)NC(NC4=CC=C(C=C4)OC(C)(C)C(O)=O)=O
Molecular Weight (g/mol) 709.57
Formula Weight 709.57
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.