missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LPPR4 Monoclonal Antibody (368701)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561218

Product Code. 30535605

  • £315.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Phospholipid-mediated signalling on neurons provokes diverse responses such as neurogenesis, pattern formation and neurite remodelling. A recently uncovered novel set of molecules in the mammalian brain, named plasticity-related genes (PRGs) mediate lipid phosphate phosphatase activity and provide evidence for their involvement in mechanisms of neuronal plasticity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LPPR4
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q7Z2D5
PLPPR4
E. coli-derived recombinant human PRG-1/LPPR4, Ser285-Asp715.
100 μg
Primary
Human
Antibody
IgG2b
Western Blot
368701
Unconjugated
PLPPR4
A330086D10; Brain-specific phosphatidic acid phosphatase-like protein 1; D3Bwg0562e; Kiaa0455; Lipid phosphate phosphatase-related protein type 4; LPPR4; LPR4; mKIAA0455; phospholipid phosphatase related 4; phospholipid phosphatase-related protein type 4; Php1; plasticity related gene 1; plasticity related gene-1; plasticity-related gene 1 protein; PLPPR4; Prg1; Prg-1; RP4-788L13.1
Mouse
Protein A/G
RUO
9890
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.