missing translation for 'onlineSavingsMsg'
Learn More

LPPR3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£175.00 - £401.00

Specifications

Antigen LPPR3
Dilution Western Blot, ELISA
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18398773
View Documents
Novus Biologicals
NBP3-11908-25UG
25 μg
£175.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18398253
View Documents
Novus Biologicals
NBP3-11908-100UG
100 μg
£401.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LPPR3 Polyclonal antibody specifically detects LPPR3 in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

LPPR3
Western Blot, ELISA
Unconjugated
Rabbit
Human
EC 3.1.3, EC 3.1.3.4, Lipid Phosphate Phosphatase-Related Protein Type 3, LPR3, PAP-2-Like Protein 2, PHP2, Plasticity-Related Gene 2 Protein, PRG2, PRG-2
Synthetic peptide derived from the LPPR3 protein (Uniprot: Q6T4P5)
Primary
Store at -70C. Avoid freeze-thaw cycles.
Western Blot, ELISA
Polyclonal
Purified
RUO
PBS
79948
IgG
Antigen Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.