missing translation for 'onlineSavingsMsg'
Learn More

LPPR2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18361872 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361872 100 μg 100µg
18390193 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18361872 Supplier Novus Biologicals Supplier No. NBP311907100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LPPR2 Polyclonal antibody specifically detects LPPR2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPPR2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias DKFZp761E1121, EC 3.1.3.4, FLJ13055, lipid phosphate phosphatase-related protein type 2, plasticity related gene 4, Plasticity-related gene 4 protein, PRG4, PRG-4
Host Species Rabbit
Immunogen Synthetic peptide derived from the LPPR2 protein (Uniprot: Q96GM1)
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64748
Target Species Human
Content And Storage Store at -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.