missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LPP Recombinant Rabbit Monoclonal Antibody (24GB05)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557446
This item is not returnable.
View return policy
Description
LPP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a member of a subfamily of LIM domain proteins that are characterized by an N-terminal proline rich region and three C-terminal LIM domains. The encoded protein localizes to the cell periphery in focal adhesions and may be involved in cell-cell adhesion and cell motility. This protein also shuttles through the nucleus and may function as a transcriptional co-activator. This gene is located at the junction of certain disease related chromosomal translocations which result in the expression of chimeric proteins that may promote tumor growth. Alternate splicing results in multiple transcript variants.
Specifications
| LPP | |
| Recombinant Monoclonal | |
| Unconjugated | |
| LPP | |
| 9430020K16Rik; AA959454; AU024130; B130055L10Rik; C79715; D630048H16; I79_000864; LIM domain containing preferred translocation partner in lipoma; LIM domain-containing preferred translocation partner in lipoma; LIM protein; lipoma preferred partner; Lipoma-preferred partner; Lipoma-preferred partner homolog; Lipoma-preferred-partner; LPP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 210126, 288010, 4026 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB05 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5XI07, Q8BFW7, Q93052 | |
| LPP | |
| A synthesized peptide derived from human LPP (aa 1-64). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction