missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25045-1-AP-20UL
This item is not returnable.
View return policy
Description
Mitotic kinesin required for chromosome passenger complex (CPC)-mediated cytokinesis. Following phosphorylation by PLK1, involved in recruitment of PLK1 to the central spindle. Interacts with guanosine triphosphate (GTP)-bound forms of RAB6A and RAB6B. May act as a motor required for the retrograde RAB6 regulated transport of Golgi membranes and associated vesicles along microtubules. Has a microtubule plus end-directed motility.Specifications
| LPP | |
| Polyclonal | |
| Unconjugated | |
| LPP | |
| Lipoma preferred partner, LPP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 210126, 4026 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BFW7, Q93052 | |
| LPP | |
| LPP Fusion Protein Ag19205 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction