missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPP Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-46145
This item is not returnable.
View return policy
Description
LPP Polyclonal specifically detects LPP in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| LPP | |
| Polyclonal | |
| Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_005569.1 | |
| LPP | |
| Peptide with sequence C-RNDSDPTYGQQGHP corresponding to internal region according to NP_005569.1. | |
| 0.1 mg | |
| Primary | |
| This antibody recognizes and epitope within the internal region. Cross reacts with Human. Expected from sequence similarity: Dog and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| DKFZp779O0231, FLJ30652, FLJ41512, LIM domain containing preferred translocation partner in lipoma, LIM domain-containing preferred translocation partner in lipoma, LIM protein, lipoma-preferred partner | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4026 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction