missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LPO Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115521
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total LPO.
This gene encodes an oxidoreductase secreted from salivary, mammary, and other mucosal glands that functions as a natural antibacterial agent. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| LPO | |
| Polyclonal | |
| Unconjugated | |
| LPO | |
| 5830499B15Rik; airway lactoperoxidase; EC 1.11.17 antibody; lactoperoxidase; lactoperoxydase; Lpo; LPO antibody; MGC129990; MGC129991; Salivary peroxidase; SAPX; SAPX antibody; SPO; SPO antibody | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 287610, 4025, 76113 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P22079 | |
| LPO | |
| A synthesized peptide derived from human LPO(Accession P22079), corresponding to amino acid residues H543-L593. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction