missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LPIN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556496
This item is not returnable.
View return policy
Description
Immunogen sequence: LATSPIPTED QFFKDIDTPL VKSGGDETPS QSSDISHVLE TETIFTPSSV KKKKRRRKKY KQDSKKEEQA ASAAAEDTCD VGVSSDDDKG AQAARGSSNA SLKEEECKEP LLFHSGDHYP LSDGDWSP Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
Plays important roles in controlling the metabolism of fatty acids at differents levels. Acts as a magnesium-dependent phosphatidate phosphatase enzyme which catalyzes the conversion of phosphatidic acid to diacylglycerol during triglyceride, phosphatidylcholine and phosphatidylethanolamine biosynthesis in the reticulum endoplasmic membrane. Acts also as a nuclear transcriptional coactivator for PPARGC1A to modulate lipid metabolism.
Specifications
| LPIN2 | |
| Polyclonal | |
| Unconjugated | |
| LPIN2 | |
| 2610511G02Rik; AI481352; AW742896; KIAA0249; lipin 2; lipin-2; Lpin2; Phosphatidate phosphatase LPIN2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9663 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92539 | |
| LPIN2 | |
| Recombinant protein corresponding to Human LPIN2. Recombinant protein control fragment (Product #RP-90709). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction