missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LPIN1 Recombinant Rabbit Monoclonal Antibody (4Y2U3)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA538057

Product Code. 17236933

  • £464.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Samples: HeLa, HepG2, NIH/3T3, RD, Mouse testis, Rat testis Immunogen sequence: MHLATSPILS EGASRMECQL KRGSVDRMRG LDPSTPAQVI APSETPSSSS VVKKRRKRRR KSQLDSLKRD DNMNTSEDED MFPIEMSSDE AMELLESSRT.

The Lipin family of nuclear proteins contains three members: Lipin-1, Lipin-2 and Lipin-3, all of which contain a nuclear signal sequence, a highly conserved amino-terminal (NLIP) domain and a carboxy-terminal (CLIP) domain. LPIN1 (Lipin-1) is crucial for normal adipose tissue development and metabolism. LPIN1 selectively activates a subset of PGC-1A; target pathways, including fatty acid oxidation and mitochondrial oxidative phosphorylation by inducing expression of the nuclear receptor PPARA. LPIN1 also inactivates the lipogenic program and suppresses circulating lipid levels. An abundance of LPIN1 promotes fat accumulation and insulin sensitivity, whereas a deficiency in LPIN1 may deter normal adipose tissue development, resulting in insulin resistance and lipodystrophy, a heterogeneous group of disorders characterized by loss of body fat, fatty liver, hypertriglyceridemia and insulin resistance.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LPIN1
Recombinant Monoclonal
0.27 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q14693, Q91ZP3
LPIN1
Recombinant fusion protein containing a sequence corresponding to amino acids 101-200 of human Lipin 1 (Q14693).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot
4Y2U3
Unconjugated
LPIN1
4631420P06; DKFZp781P1796; Fatty liver dystrophy protein; Fld; KIAA0188; lipin 1; Lipin1; lipin-1; LPIN1; mKIAA0188; PAP1; Phosphatidate phosphatase LPIN1
Rabbit
Affinity chromatography
RUO
14245, 23175, 313977
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.