missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LPIN1 Monoclonal Antibody (3D9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521419
This item is not returnable.
View return policy
Description
Peptide Sequence: EPFYAAFGNR PADVYSYKQV GVSLNRIFTV NPKGELVQEH AKTNISSYVR LCEVVDHVFP LLKRSHSSDF PCSDTFSNFT FWREPLPPFE NQDIHSASA.
The Lipin family of nuclear proteins contains three members: Lipin-1, Lipin-2 and Lipin-3, all of which contain a nuclear signal sequence, a highly conserved amino-terminal (NLIP) domain and a carboxy-terminal (CLIP) domain. LPIN1 (Lipin-1) is crucial for normal adipose tissue development and metabolism. LPIN1 selectively activates a subset of PGC-1A; target pathways, including fatty acid oxidation and mitochondrial oxidative phosphorylation by inducing expression of the nuclear receptor PPARA. LPIN1 also inactivates the lipogenic program and suppresses circulating lipid levels. An abundance of LPIN1 promotes fat accumulation and insulin sensitivity, whereas a deficiency in LPIN1 may deter normal adipose tissue development, resulting in insulin resistance and lipodystrophy, a heterogeneous group of disorders characterized by loss of body fat, fatty liver, hypertriglyceridemia and insulin resistance.
Specifications
| LPIN1 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q14693 | |
| LPIN1 | |
| LPIN1 (NP_663731.1, 792 a.a. ~ 890 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3D9 | |
| Unconjugated | |
| LPIN1 | |
| 4631420P06; DKFZp781P1796; Fatty liver dystrophy protein; Fld; KIAA0188; lipin 1; Lipin1; lipin-1; LPIN1; mKIAA0188; PAP1; Phosphatidate phosphatase LPIN1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23175 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction