missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LPHN3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA533908

Product Code. 15362501

  • £441.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Mouse, Rat, Bat, Elephant, Panda, Horse, Rabbit, Opossum, Turkey, Chicken, Lizard (100%).

Plays a role in cell-cell adhesion and neuron guidance via its interactions with FLRT2 and FLRT3 that are expressed at the surface of adjacent cells. Plays a role in the development of glutamatergic synapses in the cortex. Important in determining the connectivity rates between the principal neurons in the cortex.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LPHN3
Polyclonal
Unconjugated
ADGRL3
5430402I23Rik; ADGRL3; Adhesion G protein-coupled receptor L3; calcium-independent alpha-latrotoxin receptor 3; calcium-independent alpha-latrotoxin receptor homolog 3; Cirl3; CIRL-3; Cl3; D130075K09Rik; Gm1379; Kiaa0768; latrophilin 3; latrophilin homolog 3 (cow); latrophilin-3; LEC3; lectomedin 3; lectomedin-3; Lph3; LPHN3; mKIAA0768
Rabbit
Antigen affinity chromatography
RUO
100056929, 100338255, 170641, 23284, 319387, 422616
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q80TS3, Q9HAR2, Q9Z173
ADGRL3
Synthetic 19 amino acid peptide from C-terminus of human LPHN3.
50 μg
Primary
Human, Mouse, Rat, Rabbit, Monkey, Horse, Chicken, Bat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.