missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ LPDE Safety Wash Bottles with Leakproof Venting Valve

Product Code. 10431834 Shop All Bürkle Products
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Unlabeled
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10431834

Brand: Buerkle™ 03102055

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The venting valve ensures that the excess pressure can escape and dripping is prevented. With leakproof venting valve, contents cannot leak when bottle has fallen over as with wash bottles with ball valve or venting hole.

  • Safe filling due to wide-necked opening 39 mm neck diameter
  • Permanent print according to GHS with substance name in German, English, French and Spanish, GHS pictograms and CAS number
  • LDPE yellow transparent with light protection
  • Contents 500 ml
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Color Transparent Yellow
Material LDPE
Closure Color Red
Closure Included Yes
Product Type Safety Wash Bottle
Chemical Label Isopropanol
Closure Type Screw Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.