missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPCAT4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£111.18 - £274.38
Specifications
| Antigen | LPCAT4 |
|---|---|
| Concentration | 0.18 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Members of the 1-acylglycerol-3-phosphate O-acyltransferase (EC 2.3.1.51) family, such as AGPAT7, catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA), a precursor in the biosynthesis of all glycerolipids. Both LPA and PA are involved in signal transduction.Specifications
| LPCAT4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| Q643R3, Q6NVG1 | |
| 254531, 296048, 99010 | |
| LPCAT4 Fusion Protein Ag12281 | |
| Primary | |
| -20°C | |
| LPCAT4 |
| 0.18 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 1 AGP acyltransferase 7, 1 AGPAT 7, Acyltransferase like 3, AGPAT7, AYTL3, LPAAT eta, LPCAT4, LPEAT2 | |
| Lpcat4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title