missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPCAT3 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £393.00
Specifications
| Antigen | LPCAT3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18324796
|
Novus Biologicals
NBP3-04752-20UL |
20 μL |
£171.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18323235
|
Novus Biologicals
NBP3-04752-100UL |
100 μL |
£393.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
LPCAT3 Polyclonal specifically detects LPCAT3 in Mouse, Rat samples. It is validated for Western Blot.Spezifikation
| LPCAT3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 10162 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| 1-acylglycerophosphocholine O-acyltransferase, 1-acylglycerophosphoserine O-acyltransferase, C3F, EC 2.3.1.-, EC 2.3.1.23, EC 2.3.1.n6, LPCAT, LPLAT 5, Lyso-PC acyltransferase, Lyso-PC acyltransferase 3, Lysophosphatidylcholine acyltransferase, lysophosphatidylcholine acyltransferase 3LPSAT, Lysophosphatidylserine acyltransferase, lysophospholipid acyltransferase 5, Lyso-PS acyltransferase, MBOAT5OACT5, membrane bound O-acyltransferase domain containing 5, Membrane-bound O-acyltransferase domain-containing protein 5, nessy, O-acyltransferase (membrane bound) domain containing 5, O-acyltransferase domain-containing protein 5, putative protein similar to nessy | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 160-370 of human LPCAT3 (NP_005759.4). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts