missing translation for 'onlineSavingsMsg'
Learn More

LPCAT2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18382186 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382186 100 μL 100µL
18312027 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18382186 Supplier Novus Biologicals Supplier No. NBP304756100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LPCAT2 Polyclonal specifically detects LPCAT2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPCAT2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias 1-alkylglycerophosphocholine O-acetyltransferase, Acetyl-CoA:lyso-PAF acetyltransferase, Acetyl-CoA:lyso-platelet-activating factor acetyltransferase, acyl-CoA:lysophosphatidylcholine acyltransferase 2, acyltransferase like 1, Acyltransferase-like 1, AYTL1LysoPAFAT, DKFZp686H22112, EC 2.3.1, EC 2.3.1.-, EC 2.3.1.23, EC 2.3.1.67,1-acylglycerophosphocholine O-acyltransferase, FLJ20481, LPC acyltransferase 2, LPCAT1, LPCAT-2, Lyso-PAF acetyltransferase, LysoPC acyltransferase 2, lysophosphatidylcholine acyltransferase 2, lyso-platelet-activating factor (PAF) acetyltransferase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-544 of human LPCAT2 (NP_060309.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54947
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.