missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPCAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16112-1-AP-150UL
This item is not returnable.
View return policy
Description
Lysophosphatidylcholine (LPC) acyltransferase (LPCAT; EC 2.3.1.23) catalyzes the conversion of LPC to phosphatidylcholine (PC) in the remodeling pathway of PC biosynthesis (Nakanishi et al., 2006Specifications
| LPCAT1 | |
| Polyclonal | |
| Unconjugated | |
| LPCAT1 | |
| Acyltransferase like 2, AYTL2, LPC acyltransferase 1, lpcat, LPCAT 1, LPCAT1, Lyso PAF acetyltransferase, LysoPAFAT, LysoPC acyltransferase 1, PFAAP3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 210992, 361467, 79888 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q1HAQ0, Q3TFD2, Q8NF37 | |
| Lpcat1 | |
| LPCAT1 Fusion Protein Ag9035 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction