missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LPCAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582797
This item is not returnable.
View return policy
Description
Immunogen sequence: GVSVTDYTFE DCQLALAEGQ LRLPADTCLL EFARLVRGLG LKPEKLEKDL DRYSERARMK GGEKIGIAEF AASLEVPVSD LLEDMFSLFD ESGSGEVDLR ECVVALSV Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 89%.
PCAT1 acyltransferase (LPCAT; EC 2.3.1.23) catalyzes the conversion of LPC to hosphatidylcholine (PC) in the remodeling pathway of PC biosynthesis.
Specifications
| LPCAT1 | |
| Polyclonal | |
| Unconjugated | |
| LPCAT1 | |
| 1-acylglycerophosphocholine O-acyltransferase; 1-alkylglycerophosphocholine O-acetyltransferase; 2900035H07Rik; acetyl-CoA:lyso-PAF acetyltransferase; Acetyl-CoA:lyso-platelet-activating factor acetyltransferase; acyl-CoA:lysophosphatidylcholin; acyl-CoA:lysophosphatidylcholine acyltransferase 1; acyltransferase like 2; acyltransferase-like 2; AGPAT10; AGPAT9; Aytl2; BB137372; BC005662; C87117; LPC acyltransferase 1; LPCAT; LPCAT1; LPCAT-1; lyso-PAF acetyltransferase; lysoPAFAT; lysoPC acyltransferase 1; lysophosphatidylcholine acyltransferase 1; mLPCAT1; PFAAP3; Phosphonoformate immuno-associated protein 3; rd11; regulated by phosphonoformate; RGD1311599 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79888 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NF37 | |
| LPCAT1 | |
| Recombinant protein corresponding to Human LPCAT1. Recombinant protein control fragment (Product #RP-88774). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction