missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPCAT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£218.00 - £529.00
Specifications
| Antigen | LPCAT1 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18422471
|
Novus Biologicals
NBP1-88923-25ul |
25 μL |
£218.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18243218
|
Novus Biologicals
NBP1-88923 |
0.1 mL |
£529.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LPCAT1 Polyclonal specifically detects LPCAT1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| LPCAT1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| 1-acylglycerophosphocholine O-acyltransferase, Acetyl-CoA:lyso-PAF acetyltransferase, Acetyl-CoA:lyso-platelet-activating factor acetyltransferase, acyl-CoA:lysophosphatidylcholine acyltransferase 1, acyltransferase like 2, Acyltransferase-like 2, AYTL2, EC 2.3.1.-, EC 2.3.1.23,1-alkylglycerophosphocholine O-acetyltransferase, EC 2.3.1.67, FLJ12443, FLJ41609, LPC acyltransferase 1, lpcat, LPCAT-1, Lyso-PAF acetyltransferase, lysoPAFAT, LysoPC acyltransferase 1, lysophosphatidylcholine acyltransferase 1, PFAAP3, Phosphonoformate immuno-associated protein 3, regulated by phosphonoformate | |
| LPCAT1 | |
| IgG | |
| Affinity Purified | |
| Specificity of human LPCAT1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 79888 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GVSVTDYTFEDCQLALAEGQLRLPADTCLLEFARLVRGLGLKPEKLEKDLDRYSERARMKGGEKIGIAEFAASLEVPVSDLLEDMFSLFDESGSGEVDLRECVVALSV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title