missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LPAR6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115705
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total P2RY5.
The protein encoded by this gene belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. This gene aligns with an internal intron of the retinoblastoma susceptibility gene in the reverse orientation. Alternative splicing results in multiple transcript variants.
Specifications
| LPAR6 | |
| Polyclonal | |
| Unconjugated | |
| LPAR6 | |
| 2610302I02Rik; ARWH1; G-protein coupled purinergic receptor P2Y5; HYPT8; LAH3; LPA receptor 6; LPA-6; LPA6 Receptor; LPAR6; Lysophosphatidic acid receptor 6; oleoyl-L-alpha-lysophosphatidic acid recepto-like protein; oleoyl-L-alpha-lysophosphatidic acid receptor; P2ry5; P2Y purinoceptor 5; P2Y5; purinergic receptor (family A group 5); purinergic receptor 5; purinergic receptor P2Y G protein-coupled protein 5; purinergic receptor P2Y, G-protein coupled, 5; RB intron encoded G-protein coupled receptor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10161, 67168, 691774 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P43657, Q4G072, Q8BMC0 | |
| LPAR6 | |
| A synthesized peptide derived from human LPAR6(Accession P43657), corresponding to amino acid residues S103-F153. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction