missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LPAR3 Polyclonal Antibody
Description
Due to low expression of EDG receptors, it is recommended to use Femto Signal Substrate for Western blot. A suggested positive control for this product is RH7777 cells transfected with EDG7.
EDG7 is a member of the G protein-coupled receptor family, as well as the EDG family of proteins. This protein functions as a cellular receptor for lysophosphatidic acid and mediates lysophosphatidic acid-evoked calcium mobilization. This receptor couples predominantly to G(q/11) alpha proteins.EDG7 has been reported in human prostate, as well as rodent brain, kidney, lung, placenta, prostate, and testis. ESTs have been isolated from a normal prostate library, as well as cancer libraries of the head/neck, ovary, and thyroid.
Specifications
Specifications
| Antigen | LPAR3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | Lpar3 |
| Gene Accession No. | Q9UBY5 |
| Gene Alias | calcium-mobilizing lysophosphatidic acid receptor LP-A3; Edg7; Edg-7; endothelial cell differentiation gene 7; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 7; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 7; GPCR; HOFNH30; LPA receptor 3; LPA receptor EDG7; lpA3; LP-A3; LPA-3; LPA3 Receptor; Lpar3; Lysophosphatidic acid receptor 3; lysophosphatidic acid receptor Edg-7; putative G protein-coupled receptor snGPCR32; RP4-678I3; snGPCR32 |
| Gene Symbols | Lpar3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?