missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LPAR3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA533496

Product Code. 15354751

  • £439.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LPAR3 Polyclonal Antibody for IHC (P)

EDG7 is a member of the G protein-coupled receptor family, as well as the EDG family of proteins. This protein functions as a cellular receptor for lysophosphatidic acid and mediates lysophosphatidic acid-evoked calcium mobilization. This receptor couples predominantly to G(q/11) alpha proteins.EDG7 has been reported in human prostate, as well as rodent brain, kidney, lung, placenta, prostate, and testis. ESTs have been isolated from a normal prostate library, as well as cancer libraries of the head/neck, ovary, and thyroid.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LPAR3
Polyclonal
Unconjugated
Lpar3
calcium-mobilizing lysophosphatidic acid receptor LP-A3; Edg7; Edg-7; endothelial cell differentiation gene 7; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 7; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 7; GPCR; HOFNH30; LPA receptor 3; LPA receptor EDG7; lpA3; LP-A3; LPA-3; LPA3 Receptor; Lpar3; Lysophosphatidic acid receptor 3; lysophosphatidic acid receptor Edg-7; putative G protein-coupled receptor snGPCR32; RP4-678I3; snGPCR32
Rabbit
Antigen Affinity Chromatography
RUO
100052775, 100759265, 23566, 490187, 532829, 65086, 66025
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q8K5E0, Q9EQ31, Q9UBY5
Lpar3
Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human EDG7.
50 μg
Primary
Human, Mouse, Rat, Canine, Monkey, Bovine, Hamster, Horse, Bat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.