missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LPAR2/LPA2/EDG-4 Polyclonal specifically detects LPAR2/LPA2/EDG-4 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | LPAR2/LPA2/EDG-4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | EDG-4, EDG4FLJ93869, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor4,LPA receptor EDG4, G protein-coupled receptor, LPA-2, LPA2LPA receptor 2, lysophosphatidic acid receptor 2, lysophosphatidic acid receptor EDG4, Lysophosphatidic acid receptor Edg-4 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 250-350 of human LPAR2/LPA2/EDG-4 (NP_004711.2). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?