missing translation for 'onlineSavingsMsg'
Learn More

LPAR2/LPA2/EDG-4 Antibody [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30763916 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763916 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763916 Supplier Novus Biologicals Supplier No. AF6396JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

LPAR2/LPA2/EDG-4 Polyclonal antibody specifically detects LPAR2/LPA2/EDG-4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPAR2/LPA2/EDG-4
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias EDG-4, EDG4FLJ93869, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor4,LPA receptor EDG4, G protein-coupled receptor, LPA-2, LPA2LPA receptor 2, lysophosphatidic acid receptor 2, lysophosphatidic acid receptor EDG4, Lysophosphatidic acid receptor Edg-4
Host Species Sheep
Immunogen E. coli-derived recombinant human LPAR2, Met1-Val33, His91-Gly103, Ser168-Ser188, Asp264-Tyr279, Accession # Q9HBW0
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, GPCR, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9170
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.