missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LPAR2/LPA2/EDG-4 Polyclonal antibody specifically detects LPAR2/LPA2/EDG-4 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | LPAR2/LPA2/EDG-4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EDG-4, EDG4FLJ93869, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor4,LPA receptor EDG4, G protein-coupled receptor, LPA-2, LPA2LPA receptor 2, lysophosphatidic acid receptor 2, lysophosphatidic acid receptor EDG4, Lysophosphatidic acid receptor Edg-4 |
| Host Species | Sheep |
| Immunogen | E. coli-derived recombinant human LPAR2, Met1-Val33, His91-Gly103, Ser168-Ser188, Asp264-Tyr279, Accession # Q9HBW0 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?