missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LPAR1/LPA1/EDG-2 Monoclonal antibody specifically detects LPAR1/LPA1/EDG-2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | LPAR1/LPA1/EDG-2 |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | OTI1G6 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | edg-2, EDG2Lysophosphatidic acid receptor Edg-2, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor2,, Gpcr26, GPR26, LPA receptor 1, LPA-1, LPA1vzg-1, lysophosphatidic acid receptor 1, Mrec1.3, rec.1.3, ventricular zone gene 1, VZG1 |
| Host Species | Mouse |
| Immunogen | Full length recombinant protein of human LPAR1/LPA1/EDG-2 (NP_001392) produced in HEK293T cells. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?