missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPAR1/LPA1/EDG-2 Antibody (1020714) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB99631W
This item is not returnable.
View return policy
Description
LPAR1/LPA1/EDG-2 Monoclonal antibody specifically detects LPAR1/LPA1/EDG-2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| LPAR1/LPA1/EDG-2 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 1020714 | |
| Flow Cytometry, CyTOF-ready | |
| edg-2, EDG2Lysophosphatidic acid receptor Edg-2, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor2,, Gpcr26, GPR26, LPA receptor 1, LPA-1, LPA1vzg-1, lysophosphatidic acid receptor 1, Mrec1.3, rec.1.3, ventricular zone gene 1, VZG1 | |
| E. coli-derived recombinant human LPAR1/LPA1/EDG-2, Met1-Lys50, Accession # Q92633 | |
| 0.1 mL | |
| Apoptosis, GPCR, Signal Transduction | |
| 1902 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction