missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPA Polyclonal Antibody

LPA Polyclonal Antibody
Brand: Thermo Scientific PA563883
This item is not returnable.
View return policy
Description
LPA Polyclonal Antibody for IHC
Specifications
| LPA | |
| Polyclonal | |
| Unconjugated | |
| LPA | |
| AK38; antiangiogenic AK38 protein; apo(a); APOA; apolipoprotein(a); lipoprotein(a); lipoprotein, Lp(a); LP; lp(a); LPA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4018 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P08519 | |
| LPA | |
| Recombinant protein corresponding to Human LPA. Recombinant protein control fragment (Product #RP-103900). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction