missing translation for 'onlineSavingsMsg'
Learn More

Lp-PLA2-IN-1, MedChemExpress

Product Code. 16487888
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487888 10 mg 10mg
16453837 50 mg 50mg
16427706 100 mg 100mg
16420358 5 mg 5mg
16401859 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16487888 Supplier Medchem Express Supplier No. HY19757

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lp-PLA2-IN-1 is a potent Lipoprotein-associated phospholipase A2 (Lp-PLA2) inhibitor. Lp-PLA2-IN-1 has the potential for atherosclerosis, Alzheimer's disease research.

Lp-PLA2-IN-1 is a potent Lipoprotein-associated phospholipase A2 (Lp-PLA2) inhibitor. Lp-PLA2-IN-1 has the potential for atherosclerosis, Alzheimer's disease research.

Specifications

Chemical Name or Material Lp-PLA2-IN-1
CAS 1420367-28-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H17F5N4O3
Quantity 10 mg
Solubility Information Ethanol : 25 mg/mL (53.38 mM; Need ultrasonic)
SMILES CN1C(N2CCC1)=CC(OCC3=CC(F)=C(OC4=CC=C(C(F)(F)F)N=C4)C(F)=C3)=NC2=O
Molecular Weight (g/mol) 468.38
Formula Weight 468.38
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.