missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LP 99

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
LP 99 is a selective BRD7 and BRD9 inhibitor (Kd values are 99 and 909 nM, respectively) that exhibits selectivity for BRD7/9 over a panel of 48 bromodomains (at 10 μM). LP 99 inhibits interactions with histones H3.3 and H4 at low micromolar concentrations, and decreases the level of IL-6 secreted from LPS-stimulated THP-1 cells, in vitro. A negative control for LP 99, ent-LP 99 (Cat.No. 5497), is also available.
Specifications
Specifications
| Chemical Name or Material | N-[(2R,3S)-2-(4-Chlorophenyl)-1-(1,2-dihydro-1,4-dimethyl-2-oxo-7-quinolinyl)-6-oxo-3-piperidinyl]-2-methyl-1-propanesulfonamide |
| CAS | 1808951-93-0 |
| Quantity | 25 mg |
| Molecular Formula | C26H30ClN3O4S |
| Formula Weight | 516.05 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction