missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LOXL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550390
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total LOXL4 protein.
This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.
Specifications
| LOXL4 | |
| Polyclonal | |
| Unconjugated | |
| LOXL4 | |
| 4833426I20Rik; FLJ21889; Loxc; LOXL4; Lysyl oxidase homolog 4; lysyl oxidase like 4; lysyl oxidase related C; lysyl oxidase-like 4; lysyl oxidase-like 4 pseudogene; lysyl oxidase-like protein 4; Lysyl oxidase-related protein C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84171 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q96JB6 | |
| LOXL4 | |
| Full length fusion protein of Human LOXL4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction