Learn More
Invitrogen™ LOXL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587701
Description
Immunogen sequence: RQAPSLPLPG RVGSDTVRGQ ARHPFGFGQV PDNWREVAVG DSTGMARART SVSQQRHGGS ASSVSASAFA STYRQQPSYP QQFPYPQAPF VSQYENYDPA SRTYDQGFVY YRPAGGGVGA GAAAVASAGV IYPYQPRARY EEYGGGEELP EYPPQGFYPA PERPYVPPPP PPPDGLDRRY SHSLYSEGTP GFEQAYPDPG PEAAQAHGGD PRLGWYPPYA NPPPEAYGPP RALEPPYLPV RSSDTPPPGG ERNGAQQGRL SVGSVYRPNQ NGRGLP; Positive Samples: HeLa, A375, A-549, BT-474, HT-1080, Mouse lung, Mouse heart, Rat heart; Cellular Location: extracellular space.
This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.
Specifications
| LOXL1 | |
| Polyclonal | |
| Unconjugated | |
| Loxl1 | |
| cross-linking enzyme; LOL; Lox2; LOXL; LOXL1; Lysyl oxidase 2; lysyl oxidase homolog 1; lysyl oxidase like 1; lysyl oxidase-like 1; lysyl oxidase-like protein 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16949 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.05 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P97873 | |
| Loxl1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.