missing translation for 'onlineSavingsMsg'
Learn More

Loxiglumide, MedChemExpress

Product Code. 16478738
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478738 10 mg 10mg
16474647 50 mg 50mg
16428856 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478738 Supplier Medchem Express Supplier No. HYB2154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Loxiglumide is a cholecystokinin (CCK-1) receptor antagonist.

Loxiglumide is a cholecystokinin (CCK-1) receptor antagonist.

Specifications

Chemical Name or Material Loxiglumide
CAS 107097-80-3
Color White
Purity Grade Notes Research
Molecular Formula C21H30Cl2N2O5
Quantity 10 mg
Synonym CR-1505
Solubility Information DMSO : ≥ 100 mg/mL (216.74 mM)
SMILES O=C(O)CCC(NC(C1=CC=C(Cl)C(Cl)=C1)=O)C(N(CCCOC)CCCCC)=O
Molecular Weight (g/mol) 461.38
Formula Weight 461.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.