missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LOX Recombinant Rabbit Monoclonal Antibody (23GB920), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | LOX |
|---|---|
| Clone | 23GB920 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089674
![]() |
Invitrogen™
MA553132 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088986
|
Invitrogen™
MA553133 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LOX Recombinant Monoclonal antibody specifically detects LOX in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| LOX | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P16636, P28300, P28301 | |
| 16948, 24914, 4015 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB920 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI893619; cross-linking enzyme; H-rev142; LOX; Lysyl oxidase; Lysyl oxidase (an H-rev gene with its expression down-regulated in HRAS-transformed rat 208F fibroblasts); MGC105112; protein-lysine 6-oxidase; Protein-lysine 6-oxidase, long form; Protein-lysine 6-oxidase, short form; Ras excision protein; ras recision; rrg; Rrg1; TSC-160 | |
| A synthetic peptide of human LOX (350-417AA). | |
| Primary | |
| -20°C | |
| LOX |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title