missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOX Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17958-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is an extracellular copper enzyme that initiates the crosslinking of collagens and elastin. The enzyme catalyzes oxidative deamination of the epsilon-amino group in certain lysine and hydroxylysine residues of collagens and lysine residues of elastin. In addition to crosslinking extracellular matrix proteins, the encoded protein may have a role in tumor suppression.Specifications
| LOX | |
| Polyclonal | |
| Unconjugated | |
| LOX | |
| LOX, lysyl oxidase, Protein lysine 6 oxidase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16948, 4015 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P28300, P28301 | |
| LOX | |
| LOX Fusion Protein Ag12479 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction